J.T.Baker® BAKERBOND Speedisk®, High Throughput SPE Polymer Columns
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKR8109-09 BAKR8108-04 BAKR8108-09 BAKR8109-06
CAJT8109-6PK 2040.95 CAD
CAJT8109-6 CAJT8109-9 CAJT8108-04 CAJT8108-06 CAJT8108-09 CAJT81096 CAJT81114
J.T.Baker® BAKERBOND Speedisk®, High Throughput SPE Polymer Columns
Chromatography Columns
These columns feature BAKERBOND® DVB polymer resins, which are the product of ultra-clean polymer microparticle technology.
These resin particles have a large surface area, are highly rigid, and are stable over pH range 1 to 14. These columns are recommended when advanced detection methods will be used, and offer the unique selectivity of a polymer. The table below lists the typical use for each column type.