Corning® Thermowell® GOLD 96-Well Clear Polypropylene PCR Microplate, Corning
Supplier: Corning
Corning®
3758
89090-032CS 822.56 CAD
89090-032
Corning® Thermowell® GOLD 96-Well Clear Polypropylene PCR Microplate, Corning
Microplates PCR Plates
These half skirted 96 well PCR microplates feature a rigid construction that is ideal for automation.
  • Clear medical grade PP is suitable for RT-PCR
  • Uniform wall thinness assures efficient heat transfer while minimizing well-to-well variability
  • 100% tested for well integrity and DNase- and RNase-free
  • Nonsterile, without lids
  • With two generic bar code labels per plate