Soxhlet Extraction Apparatus, Chemglass
Supplier: Chemglass
CG-1368-01 CG-1368-02 CG-1368-03 CG-1369-01 CG-1369-02 CG-1369-03
80068-152EA 1135.8 CAD
80068-152 80068-154 80068-156 80068-170 80068-172 80068-174
Soxhlet Extraction Apparatus, Chemglass
Extractors Soxhlet and Soxhlet-Type Extractors
Used for continuous extraction of solids with a suitable solvent
Complete apparatus consists of an Alihn condenser, extractor, and flat or round bottom flask. Extractor uses paper or glass thimbles (see 80068-184 series). Condenser has an enlarged uptake tube to improve condensing capacity.