Falcon® Round-Bottom Polystyrene Tubes, Disposable, Corning
Supplier: Corning
352235 352052 352017 352003 352058 352027 352037 352025 352045 352001 352057 352051 352008 352054
CA21008-948CS 1621.44 CAD
CA21008-948 CA60819-138 CA60819-218 CA60819-273 CA60819-295 CA60819-331 CA60819-375 CA60819-422 CA60819-433 CA60819-499 CA60819-524 CA60819-546 CA60819-820 CA60819-310
Falcon® Round-Bottom Polystyrene Tubes, Disposable, Corning
Tubes Culture Tubes
These disposable, clear polystyrene tubes are best suited for procedures requiring high optical clarity, and thermal and chemical stability. Provide reliable containment of laboratory fluid samples.
  • Dual position snap-cap prevents sample loss with ease of handling, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • Tube dimensions and volumes are approximate
  • Widely used and referenced in laboratory protocols
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/ml (Cat. No. 352008 nonsterile)
Polyethylene snap-caps feature a unique, two-position fit: loose for aerobic culturing, tight for sealing. 1400 RCF rating (RCF claims refer to relative centrifugal force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0 °C may have lower RCF ratings).