EasyCurve™ Mop System, Contec®
Supplier: Contec
2724 EZMP0350 EZMP0300 EZSE0100 EZSE0150
89073-976EA 239.91 CAD
89073-976 CA10053-838 CA10153-828 10816-276 10816-298
EasyCurve™ Mop System, Contec®
Mops
The EasyCurve™ Mop was designed specifically for use in critical and controlled environments. It is now available with a ribbed microfiber face, ensuring smooth and easy mopping with improved particle pick-up and contamination removal.
  • All components are autoclavable; ideal for a wide range of cleaning applications in the most critical sterile environments
  • Lightweight mop head and handle significantly reduces operator fatigue
  • Single-use mop is economical and easy to install without tools
  • Curved mop and pivoting mop frame provide effortless contact with
More Product Information