VWR® Angled Foam Wall Mop
Supplier: VWR
VWR®
89012-766
89012-766CS 1148.14 CAD
89012-766
VWR® Angled Foam Wall Mop
Mops
Wedge-shaped mop head is made of knitted polyester fabric laminated to clean ester foam for excellent abrasion resistance.
  • Fully autoclavable
  • Stainless steel swivel-head frame
Ideal for cleaning ceilings and walls, and for applying disinfectants and cleaning solvents in controlled environments
Swivel-head frame is durable stainless steel. Features a capped end for maximum cleanliness and a quick-connect button for easy attachment to interchangeable handles.
Telescoping handles are available in two materials, lightweight anodized aluminum or durable stainless steel, and two lengths.
Large size allows efficient coverage of large areas, while wide leading edge and angled sides allow easy access to tight corners. Compatible with a wide range of chemicals. Autoclavable at 121°C (250°F) for 30 minutes. Suitable for ISO Class 5 (FED-STD-209D Class 100/M3.5) conditions. Fits a 30.5 cm (12") frame (not included).
Ordering information: Mop head, mop frame, and handle must all be purchased separately.