KIMBLE® KONTES® ULTRA-WARE® All-Glass Funnel/Support Assembly, 47 mm, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
953825-0000 953835-0000 953845-0000
KT953825-0000EA 1505.36 CAD
KT953825-0000 KT953835-0000 KT953845-0000
KIMBLE® KONTES® ULTRA-WARE® All-Glass Funnel/Support Assembly, 47 mm, DWK Life Sciences
Filters Membrane Filter Units
Designed for vacuum filtration of corrosive liquids for analysis of particulate contamination.
Unique base design has integral vacuum connection located above filtrate drip to prevent contamination of vacuum line with filtrate droplets.
Unit consists of a 300 ml funnel; a coarse-porosity, fritted-glass filter support base with integral vacuum connection; and a 1 L inner joint flask. An aluminum clamp connects funnel and support base. [ST] joints are 40/35.
Liquids come in contact only with borosilicate glass.