Continuous Extractor, Liquid-Liquid, Chemglass
Supplier: Chemglass
CG-1360-01
80068-146EA 1411.72 CAD
80068-146
Continuous Extractor, Liquid-Liquid, Chemglass
Extractors Liquid-Liquid Extractors
Designed for EPA priority pollutant samples where the extracting solvent is heavier than water
For use with samples of about 1L plus approximately 200mL of extracting solvent. Extractor has a [ST] 45/50 top outer joint and a [ST] 24/40 lower inner joint.
Ordering information: Accessories sold separately.