Tilting Dispensers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
759300-0100 759300-0020 759300-0040
KT759300-0015EA 843.07 CAD
KT759300-0015 KT759300-0030 KT759300-0100 KT759300-0005 KT759300-0010 KT759300-0020 KT759300-0025 KT759300-0040 KT759300-0050 KT759300-0060 KT759301-0005 KT759301-0010 KT759301-0015 KT759301-0020 KT759301-0025 KT759301-0030 KT759301-0040 KT759301-0050 KT759301-0060 KT759301-0100
Tilting Dispensers, Kimble Chase
Bottle Top Tilt Measures
All-glass apparatus with fixed volume dispensing head
A polyacetal [ST] clamp (included) secures flask and head. EPA Method SW846.
Order Now
SPECIFICATIONS
Environmentally Preferable