Chucks and Flexible Couplings, Arrow
Supplier: Arrow Engineering
JACOBS-250 JACOBS-312 JACOBS-375 NYLEX-250 NYLEX-312 NYLEX-375 D1-250 D1-312 D1-375 D1
59025-174EA 181.53 CAD
59025-174 59025-176 59025-178 59025-180 59025-182 59025-184 59025-186 59025-188 59025-190 59025-192 59025-194 59025-196 59025-198 59025-200
Chucks and Flexible Couplings, Arrow
Stirrers Stirring Tools
Adjustable Jacobs Chucks with key accept stirrer shaft diameters from 1.6–9.5mm (1/163/8")
Nylon-Bodied Keyless Chucks accept stirrer shaft diameters from 1.6–9.5mm (1/163/8"). Available in three motor shaft diameters.
Flexible Couplings provide a safe and convenient alternative for working with glass stirring shafts. Length of 10.2cm (4") allows for angular and parallel misalignment of glass shaft without danger of breakage. One end of coupling contains a corrosion-resistant standard metal collar with a stainless steel set screw for fastening to the stirrer shaft. Collar fits into a neoprene rubber sleeve body that accepts a replaceable fluoropolymer resin sleeve with set screw. Fluoropolymer resin sleeve securely holds a glass shaft of 10mm (3/8") or 19mm (3/4") diameter. Rubber sleeve body absorbs shock and provides vibration-free, quiet-running and great torsional stiffness. Corrosion-resistant metal collar, neoprene rubber and stainless steel set screw provide long life in difficult environments. Only fluoropolymer resin sleeve makes contact with glass shaft.
Available in three motor shaft diameters.
Ordering information: Flexible Couplings, complete with fluoropolymer resin sleeve and two stainless steel set screws, are available in five motor shaft diameters.