Digital 'Pocket' Refractometers and Special Scales, ATAGO®
Supplier: Atago
3820-PAL-2 3810-PAL-1 3830
12000-668EA 981.65 CAD
12000-668 47752-918 80093-902
Digital 'Pocket' Refractometers and Special Scales, ATAGO®
Refractometers Handheld Refractometers
These compact, pocket-sized refractometers allow you to take measurements anywhere with ease, indoors or outdoors
The units can take a measurement of a high temperature sample that has been heated or boiled. Rinse off samples easily by running water on the sample stage. The units eliminate the need for frequent zero setting; they only require zero setting at the start of each workday. Three models are available to suit a wide range of measurement needs.
To use, simply place a drop of sample on the prism surface and press the start key. The Brix value will be displayed in 3 seconds onMore Product Information