Variable-Speed Air-Powered Stirrers, Arrow
Supplier: Arrow Engineering
TYPE A TYPE G TYPE G-HD TYPEGG TYPE A-5 TYPE G-5
59025-100EA 1476.35 CAD
59025-100 59025-102 59025-104 59025-106 59025-108 59025-110
Variable-Speed Air-Powered Stirrers, Arrow
Stirrers Overhead Stirrers
Explosion-proof stirrers for safe mixing of all types of solvents, lacquers, and volatile chemicals
Needle valve allows user to vary mixing speed. A special muffler ensures quiet operation. A number of non-reversible models are available for high speed, general purpose, and high torque mixing applications. Model A — Low-cost, light-duty stirrer ideal for stirring all types of solvents, lacquers, oils, synthetics, and chemicals. Temperature range: ambient to 120°C (250°F). Model A-5 — Similar to Model A, but 1/2 hp motor provides more power and is recommended for larger batches.More Product Information