Purifier® Non-Ventilated PCR Enclosure, Labconco®
Supplier: Labconco
Purifier®
3908402 3747002 3952400 3952420 3747012 3747002
89000-160EA 12805.9 CAD
89000-160 82003-562 82003-568 82010-694 470203-172 89000-158
Purifier® Non-Ventilated PCR Enclosure, Labconco®
Fume Hoods and Enclosures PCR Workstation Enclosures
Provides a non-ventilated, circulation-free work area designed to reduce cross-contamination during polymerase chain reaction (PCR) experiments. The unit is constructed of UV-resistant, glacier white, dry powder epoxy-coated aluminum and steel with 0.6cm (1/4") tempered safety glass sash and sides. Angled, hinged sash features a 19.8cm (713/16") high opening for closer, more comfortable viewing than vertical sashes, and an interlocked UV protection panel that must be in place to turn on UV light but can be removed when the light is off for full access to enclosure opening.
A fluorescent light provides a bright, nonglare working area
An optional work surface is available with raised edges to contain spills. Base stands are made of durable 4.4cm (13/4") thick, tubular, glacier white epoxy-coated steel and have an adjustable height from 70–85cm (271/2–331/2") in 2.5cm (1") increments. Stands with casters feature 12.7cm (5") diameter toe-locking casters with bearings for quiet operation.
The UV light is controlled by a variable digital timer that provides continuous or timed UV light exposure in intervals of 5, 10, 15, 30, 60, 120, and 240 minutes to deactivate DNA and RNA contaminants and to ensure thorough decontamination while the enclosure is unattended. An amber LED indicator blinks for the duration of the setting.
Certifications: The 115V model is ETL and ETL-C listed. The 230V model is CE marked.
Ordering information: Manufacturer’s one-year warranty on parts and labor. Each enclosure includes a grounded, switched electrical receptacle and a 2.7m (9') three-wire cord with appropriate plug. Base stand or table and work surface (required for operation) are not included. Items designated below as REDISHIP can be shipped within 48 hours of receiving your order. An interlocked UV Protection Panel replaced the bi-fold doors.
Caution: Warning: This is a non-ventilated, circulation-free enclosure. It is not suitable for biohazardous, toxic, or volatile procedures. Because there is no exhaust system in the enclosure, continuous flame or high heat sources should not be used.