KONTES® CHROMAFLEX™ Safety/Water-Jacketed Chromatography Columns, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
420870-6010 420870-1000 420870-1010 420870-1020 420870-1200 420870-1210 420870-1220 420870-1500 420870-1510 420870-1520 420870-3000 420870-3010 420870-3020 420870-6020
KT420870-6000EA 1421.27 CAD
KT420870-6000 KT420870-6010 KT420870-1000 KT420870-1010 KT420870-1020 KT420870-1200 KT420870-1210 KT420870-1220 KT420870-1500 KT420870-1510 KT420870-1520 KT420870-3000 KT420870-3010 KT420870-3020 KT420870-6020
KONTES® CHROMAFLEX™ Safety/Water-Jacketed Chromatography Columns, Kimble Chase
Chromatography Columns
For medium pressure chromatography with either aqueous or organic mobile phases.
Flow adapters allow easy adjustment of column bed volume and offer improved column performance and chromatogram resolution. Flow adapters are constructed with PTFE eluant cannula, acetal body, PTFE/propylene O-ring seal, and 20 µm polyethylene bed support.
A PTFE bed support is required when flow adapter is used with aggressive organic solvents, PTFE bed supports include 20 µm PTFE bed support, 40 mesh PTFE support screen, and PTFE retainer ring.
These threaded borosilicate glass columns feature an acrylic water jacket that provides thermal control of temperature-sensitive chromatographic separations and acts as a safety shield.
Ordering information: Each column is supplied with a 20 µm polyethylene bed support and ¹/₄"-28 fittings.