KONTES® CHROMAFLEX™ Chromatography Columns, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
420830-1510 420830-1520 420830-3000 420830-3010 420830-3020 420830-6020 420830-1000 420830-1010 420830-1020 420830-1200 420830-1210 420830-1220 420830-1500 420830-6000 420830-6010
KT420830-1510EA 832.63 CAD
KT420830-1510 KT420830-1520 KT420830-3000 KT420830-3010 KT420830-3020 KT420830-6020 KT420830-1000 KT420830-1010 KT420830-1020 KT420830-1200 KT420830-1210 KT420830-1220 KT420830-1500 KT420830-6000 KT420830-6010
KONTES® CHROMAFLEX™ Chromatography Columns, Kimble Chase
Chromatography Columns
Ideal for medium pressure chromatography with either aqueous or organic mobile phases.
Packing reservoirs are threaded for easy mounting on columns. They are designed for use as buffer reservoirs in gravity flow chromatography or for packing gel slurry into columns (reservoir capacity matches the volume of gel required to pack the longest column in that diameter.
Flow adapters allow easy adjustment of column bed volume and offer improved column performance and chromatogram resolution. Flow adapters are constructed with PTFE eluant cannula, acetal body, PTFE/propylene O-ring seal, and 20 µm polyethylene bed support.
A PTFE bed support is required when flow adapter is used with aggressive organic solvents, PTFE bed supports include 20 µm PTFE bed support, 40 mesh PTFE support screen, and PTFE retainer ring.
These threaded columns feature PTFE shielded O-rings that form positive, leak-free seals. The solvents contact only the PTFE and borosilicate glass. The 10 cm Int.Ø columns are tapered at both ends, enabling them to utilize the end fittings of 4.8 cm Int.Ø columns. Recommended for RCRA SW-846 Methods.
Ordering information: Each column comes with a polyethylene bed support and ¹/₄"-28 PTFE end fittings.