Biomek® Tube Rack and Tube Inserts, Beckman Coulter®
Supplier: Beckman Coulter
Biomek®
373699 373698 373697 373698 373696 373661 373661 373699 373697
CABK373699CS 306.02 CAD
CABK373699 CABK373698 CABK373697 CABK373696 CABK373661 BK373699 BK373698 BK373697 BK373661
Biomek® Tube Rack and Tube Inserts, Beckman Coulter®
Racks Tube Racks
The 24-place rack allows the use of various-sized glass, polystyrene, polypropylene, and polyallomer tubes, Microfuge® centrifuge tubes, and cryogenic vials
To ensure proper tube fit, inserts must be used (sold separately).