Single Drum Spill Containment Station, Eagle Manufacturing
Supplier: Eagle Manufacturing
1660 1698 1612
56610-151EA 291.94 CAD
56610-151 56617-341 56617-347
Single Drum Spill Containment Station, Eagle Manufacturing
Drum Storage
For secondary spill containment of hazardous materials.
Blow-molded high-density polyethylene unit with 321.8L (85gal.) spill capacity is suitable for 208.2L (55gal.) drums or smaller. Use as a collection or dispensing center. Chemically-resistant to most materials. Forkliftable. Nestable for easy storage.