Fume Adsorbers, Labconco®
Supplier: Labconco
3955200 3955220 3955400 3955420 3955201 3955202 3955221 3955222 3955401 3955402 3955421 3955422
89002-422EA 5656.4 CAD
89002-422 89002-424 89002-426 89002-428 89004-796 89004-798 89004-800 89004-802 89004-804 89004-806 89004-808 89004-810
Fume Adsorbers, Labconco®
Fume Extractors
Fume adsorbers are ideal for light-duty applications involving ammonia, formaldehyde, organic solvents, and low molecular weight amines
Replacement filter utilizes 2.7kg (6lbs.) of activated carbon to adsorb vapors up to one-third of the filter weight.
Carbon filtration traps vapors, ensuring only safe, filtered air is returned to the laboratory. No outside ducting is required. A quiet, built-in motorized impeller provides face velocity up to 35fpm; models with two filters include two motorized impellers. A rear-mounted air sampling port allows testing the saturation of the carbon filter using detector tubes (not included). Units are pre-wired with a rear-mounted auxiliary electrical receptacle and a top-mounted blower switch with a circuit breaker. Portable, low-profile enclosures fit easily on countertops with overhanging cabinets. Adsorbers feature a glacier white, dry powder epoxy-coated steel housing with a perforated baffle for uniform airflow. A shatterproof, 0.95cm (3/8") thick canopy constructed of clear acrylic helps keep fumes centralized. Adsorbers are available with one or two filters for ammonia, formaldehyde or organic vapors.
Certifications: The 115V models are ETL and ETL-C listed. The 230V models are CE marked.
Ordering information: The 115V models are supplied with a 2.4m (8') three-wire power cord with a plug. The 230V models are supplied with a 2.4m (8') three-wire power cord without a plug. All models require a work surface and a base stand or cabinet. For work surfaces, base stands, cabinets, or optional accessories, contact your Avantor sales representative.