VWR® Spinring® Magnetic Stir Bars
Supplier: VWR International
58949-018 58949-020 58949-022
58949-018CS 3384.43 CAD
58949-018 58949-020 58949-022
VWR® Spinring® Magnetic Stir Bars
Stirrers Magnetic Stirring Bars
Octagonal stirring bar is contained within a removable stabilizing ring that centers it in a container and creates a deep vortex without splashing, chattering, or spinout.
  • Removable stabilizing ring keeps bar centered
  • Virtually eliminates spinout
  • No chattering noise
  • Autoclavable
Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.