SmartWall Plus™ Grids and Shelving Accessories, Metro™
Supplier: Metropolitan Wire
WG1836K3 WG1848K3 WG1860K3 WG2436K3 WG2448K3 WG3036K3 WG3048K3 GS1436K3 GS1448K3 GS1830K3 GS1836K3 GS1848K3 FGS1224K3 BH6K3 PGHK6K3
CA82023-608EA 139.48 CAD
CA82023-608 CA82023-610 CA82023-612 CA82023-614 CA82023-616 CA82023-618 CA82023-620 CA82023-622 CA82023-624 CA82023-626 CA82023-628 CA82023-630 CA82023-632 CA82023-636 CA82023-634
SmartWall Plus™ Grids and Shelving Accessories, Metro™
Graticules, Reticles and Grids
Grids and shelving accessories allow users to organize SmartWall Plus storage systems to precise specifications.
  • Grids maintain a total unit weight capacity of 114 kg (250lbs.)
Storage options can be easily repositioned along the track before securing them to the wall to maximize storage space and improve organization. Metroseal three finishes provide Microban® built-in product protection.
Ordering information: Accessory bracket kits for wire grids (82023-344) include two top brackets that attach to track and four bottom brackets that attach to wall surface. One kit is required per grid. Brackets are constructed of Type 304 stainless steel. Bracket kits for Starsys® wall cabinets (82023-334) include two top brackets and one bottom bracket. Top brackets are Type 304 stainless steel and bottom brackets are steel coated with corrosion-resistant taupe epoxy.