Glove Box Gloves, Labconco
Supplier: Labconco
5006000 5033100 5005601 5005600 5005800 5006100
15202-519EA 199.08 CAD
15202-519 32948-201 32948-203 32948-205 32948-019 32948-096 32948-110 32948-154
Glove Box Gloves, Labconco
Gloves
Neoprene straight and accordion sleeves can be used with hand-specific neoprene or latex hands.
  • Hypalon® gloves are resistant to abrasion, ozone, and oxidizing chemicals
Neoprene, butyl, and Hypalon® gloves are one-piece, seamless, and hand-specific. Butyl gloves provide high impermeability to water vapor, gases, and toxic chemicals, and are resistant to oxygenated solvents and most oxidizing chemicals.
Ordering information: Sleeves come complete with O-rings and spring. Neoprene, butyl, and Hypalon® gloves come with O-rings.