Super Erecta Shelf® Post-Type and Direct Wall Shelf Mounts, Metro™
Supplier: Metropolitan Wire
SW21C SW23C SW25C SW26C AW21C AW23C AW25C AW26C SW31C SW33C SW35C SW36C AW31C AW33C AW35C AW36C SW41C SW43C SW45C SW46C AW41C AW43C AW45C AW46C SW51C SW53C SW55C SW56C AW51C AW53C AW55C AW56C 13PDF 33PDF 54PDF 63PDF BES BCS 13PDFS 33PDFS 54PDFS 63PDFS SBCS 1WS14C 1WS18C 1WS21C 1WS24C 1WS14S 1WS18S 1WS21S 1WS24S 2WS14C 2WS18C 2WS21C 2WS24C 2WS14S 2WS18S 2WS21S 2WS24S 1WD14C 1WD18C 1WD21C 1WD24C 1WD14S 1WD18S 1WD21S 1WD24S 2WD14C 2WD18C 2WD21C 2WD24C 2WD14S 2WD18S 2WD21S 2WD24S
CA14233-016EA 544.34 CAD
CA14233-016 CA14233-018 CA14233-020 CA14233-022 CA14233-024 CA14233-026 CA14233-028 CA14233-030 CA14233-032 CA14233-034 CA14233-036 CA14233-038 CA14233-040 CA14233-042 CA14233-044 CA14233-046 CA14233-048 CA14233-050 CA14233-052 CA14233-054 CA14233-056 CA14233-058 CA14233-060 CA14233-062 CA14233-064 CA14233-066 CA14233-068 CA14233-070 CA14233-072 CA14233-074 CA14233-076 CA14233-078 CA14233-080 CA14233-082 CA14233-084 CA14233-086 CA14233-088 CA14233-090 CA14233-092 CA14233-094 CA14233-096 CA14233-098 CA14233-100 CA14233-102 CA14233-104 CA14233-106 CA14233-108 CA14233-110 CA14233-112 CA14233-114 CA14233-116 CA14233-118 CA14233-120 CA14233-122 CA14233-124 CA14233-126 CA14233-128 CA14233-130 CA14233-132 CA14233-134 CA14233-136 CA14233-138 CA14233-140 CA14233-142 CA14233-144 CA14233-146 CA14233-148 CA14233-150 CA14233-152 CA14233-154 CA14233-156 CA14233-158 CA14233-160 CA14233-162 CA14233-164 CA14233-166
Super Erecta Shelf® Post-Type and Direct Wall Shelf Mounts, Metro™
Furniture Shelves
Wall mounts for shelves are an ideal method for storing items overhead or above work surfaces.
  • All prepackaged post-type units feature a chrome-plated finish
  • Metal endcaps to cover post openings are provided
  • Use single supports at shelf ends and double supports for adjoining shelves
  • Direct wall mounts are available in chrome-plated or stainless steel finish
  • Direct wall systems have a maximum load rating of 91 kg (200 lbs.)
  • Post-type systems have a maximum load rating of 113.3 kg (250 lbs.)
Post-type wall mounts are available in prepackaged end units or mid units, which include all necessary components except wall bolts or screws. End units consist of two posts with brackets, and two single shelf supports for each shelf. Mid units consist of one post with brackets and one double shelf support for each tier. If shelving is to be the length of one shelf, order one end unit only.
Posts, brackets, and shelf supports may also be purchased separately to construct post-type wall mount systems. Each post is supplied with two end brackets. The 85.4 cm (33⁵/₈") long posts also include one intermediate bracket; the 137.2 cm (54") and 157.5 cm (62") long posts also include two intermediate brackets. Intermediate brackets can also be used to attach freestanding shelving to a wall. Post-type components are available in chrome-plated or stainless steel finish.
Direct wall mounts eliminate the need for a post; the shelf support is held by a mounting plate attached directly to the wall. This design also conserves space, as it allows the shelf and shelf support to be much closer to the wall. Each direct wall mount consists of one shelf support, one bracket, and two metal endcaps.
Brackets accommodate all types and sizes of Super Erecta® shelves (shelves sold separately).
Ordering information: If shelving is to be the length of two shelves, order one end unit and one mid unit; if three shelf lengths, order one end and two mid units and so on.