MetroMax i™ Shelves and Posts, Metro™
Supplier: Metropolitan Wire
MX1824F MX1836F MX1836G MX1872F MX2430F MX2436F MX2448F MX2472F MX2472G MX74UP MX1824F MX1836F MX1836G MX1872F MX2430F MX2436F MX2448F MX2472F MX2472G MX74UP
89369-930EA 536.2 CAD
89369-930 89369-932 89369-934 89369-936 89369-938 89369-940 89369-942 89369-944 89369-946 89369-948 CA89369-930 CA89369-932 CA89369-934 CA89369-936 CA89369-938 CA89369-940 CA89369-942 CA89369-944 CA89369-946 CA89369-948
MetroMax i™ Shelves and Posts, Metro™
Furniture Shelves
Microban® antimicrobial product protection built in to the shelf mats to inhibit stain and odor causing bacterial growth.
  • Posts feature polymer construction and are adjustable at 2.5 cm (1") increments
  • Solid mats are excellent for spill containment
Shelves consist of a shelf frame, open-grid or solid shelf mat, and four wedge connectors. Open-grid mats allow for air circulation and reduce dust and moisture buildup. Shelf mats may be removed from the shelf frame for easy cleaning.
Shelves may be adjusted in 2.5 cm (1") increments on the shelf posts (More Product Information