JS-4.2SM and JS-4.2SMA Swinging Bucket Rotors, Beckman Coulter®
Supplier: Beckman Coulter
348394 366670
CABK348394EA 25715.11 CAD
CABK348394 CABK366670
JS-4.2SM and JS-4.2SMA Swinging Bucket Rotors, Beckman Coulter®
Centrifuges Rotors
Windshielded, six-place swinging bucket rotors feature oval-shaped buckets designed specifically for blood bank applications.
  • ARIES™ Smart Balance Technology Included
  • Specific Use for Blood Bank Applications
  • Six-Place, Windshielded Swinging Bucket Rotors
  • Space for Six Blood Bags
  • Imbalance Compensation for Buckets Unbalanced by up to 100 g.
Rotors JS-4.2SMA feature ARIES™ Smart Balance technology, which provides imbalance compensation for rotors with buckets that are unbalanced by up to 100 g.
These rotor are for use in the Avanti™ J6-MI centrifuge. Rotors accommodate six blood bags. The buckets are interchangeable with microplate carriers (sold separately); each carrier holds up to three microtiter plates.
Ordering information: Rotors include six oval-shaped blood bag cups.