Digital Urine Specific Gravity Refractometer, Model UG-[alpha], ATAGO®
Supplier: Atago
3464
89009-084EA 4349.33 CAD
89009-084
Digital Urine Specific Gravity Refractometer, Model UG-[alpha], ATAGO®
Refractometers Handheld Refractometers
Model UG-[alpha] is a compact urine specific gravity refractometer with digital display
Simple operation involves placing sample onto prism and pressing start. Measurements will stream across the LCD display. The minimum scale indication displays up to the fourth decimal. The UG-[alpha] refractometer is equipped with Automatic Temperature Compensation to provide accurate measurements, regardless of environmental conditions.
Ordering information: Uses one 9V battery.