Temperature Probes
Supplier: Mettler Toledo
51343310 51343312 51302046 51300164 51300165
CA97001-698EA 1150.26 CAD
CA97001-698 CA97001-700 CA11277-306 CA11278-744 CA11278-746
Temperature Probes
Temperature Probes
These sensing elements are housed in glass or tubular stainless steel and are designed for connection to METTLER TOLEDO pH meters.
  • Glass or stainless steel housing
  • Lead wires isolated from outer probe
Probes have a maximum operating temperature of 150 °C (302 °F) and are constructed with the sensing elements and lead wires electrically isolated from the outer probe surface for maximum safety.
Ordering information: Probes are part of the InLab® product line and are supplied with quality certificate.