Buchner Filter Funnels, [ST] Joint, Ace Glass
Supplier: Ace Glass
9439-02 9439-04 9439-06 9439-08 9439-10 9439-11 9439-13 9439-15 9439-17 9439-19 9439-22 9439-24 9439-26 9439-28 9439-30 9439-40 9439-42 9439-44 9439-50 9439-52 9439-54
89074-532EA 290.86 CAD
89074-532 89074-534 89074-536 89074-538 89074-540 89074-542 89074-544 89074-546 89074-548 89074-550 89074-552 89074-554 89074-556 89074-558 89074-560 89085-094 89085-096 89085-098 89085-100 89085-102 89085-104
Buchner Filter Funnels, [ST] Joint, Ace Glass
Funnels Filtration Funnels Büchner Funnels
Buchner-type filter funnels contain an integral 40mm diameter sintered glass filter disc
Available in five different porosities. Fabricated from borosilicate glass.