VWR® Traceable® Extreme-Accuracy Digital Thermometers
Supplier: VWR International
89094-752 89094-754 89094-756 89094-758
89094-752EA 405.83 CAD
89094-752 89094-754 89094-756 89094-758
VWR® Traceable® Extreme-Accuracy Digital Thermometers
Thermometers Digital Thermometers
Absolute high-precision thermometers provide the exact readings necessary for critical requirements, quality control checks, and routine measurements.
  • Traceable
  • Temperature in °C and °F
  • Stainless steel probe
  • Continuous visual and audio alerts of temperature changes
  • One-button self-calibration
These thermometers are ideal for monitoring temperatures in water baths, cuvettes, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater
The stainless steel probe can be used in liquids, semi-More Product Information