KONTES® Stirrer Assembly, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
788000-0021 781030-0124 788010-0021
KT788000-0021EA 104.93 CAD
KT788000-0021 KT781030-0124 KT788010-0021
KONTES® Stirrer Assembly, Kimble Chase
Stirrers Stirring Shafts
Complete assembly with glass shaft and bearing
Blade is PTFE. Stirrer adapters (sold separately) permit use with stirrer motors. Assembly designed for use with flask holding a volume of 500–5000mL and having a [ST] 24/40 joint. Approximate overall dimensions are 445Lx75Wmm (17.5x3") with a 10mm shaft O.D.