Accessories for Explosion-Proof Stirrer Lightnin®
Supplier: Lightnin
829448 143729 800570 800569 809417 800568 143264 143259 143728 143191
58958-066EA 2795.1 CAD
58958-066 58958-164 58958-200 58958-222 58958-233 58958-244 58958-266 58958-277 58958-186 58958-255
Accessories for Explosion-Proof Stirrer Lightnin®
Stirrers Overhead Stirrers
The 316 Stainless Steel shafts are available in 7.9 mm (5/16") and 9.5 mm (3/8") diameters and three lengths.
The A100 impellers have traditional three-blade design for moderate pumping action and wetting dry powders
Heavy-duty ring stand clamp of cast 316 stainless steel is recommended for all models with tube suspension mounting.
Appropriate for mixing low- to high-viscosity materials. Available with 7.9 mm (5/16") bore diameter and four blade diameters.