Stands for Heidolph® Hei Torque Overhead Stirrers, Heidolph
Supplier: Heidolph NA, LLC
036300520CA 036300530CA 036300540CA
CA11009-814EA 975.23 CAD
CA11009-814 CA11009-816 CA11009-818
Stands for Heidolph® Hei Torque Overhead Stirrers, Heidolph
Retort Stands
Designed for use with the Heidolph Hei Torque series of overhead stirrers. These stands provide sturdy support for stirrers during mixing procedures.
The Universal stand S2 has a length of 700mm. The Universal stand S2 XXL has a length of 1000mm. The Telescope stand provides vertical displacement of 725–1025mm. All stands feature an H-shaped base measuring 425x420mm. Fastening clamps are not included.
Order Now
SPECIFICATIONS
Environmentally Preferable