VWR® Talon® Support Stands
Supplier: VWR International
30400030 30400032 30400035 30400031 80862603 30400034
14010-002EA 962.49 CAD
14010-002 14010-004 14010-010 58958-459 97035-508 14010-008
VWR® Talon® Support Stands
Retort Stands
Support stands are made entirely of Type 303 stainless steel and can accommodate vessels up to 45.7 cm (18") in diameter within the sturdy U-shaped base.
  • Stainless steel construction
  • Sturdy U-shaped base
  • Includes support rod
  • Fitted with oversized knobs for very secure positioning
The threaded stainless steel support rod is 16 mm (5/8") in diameter and is screwed into the support stand base
Accessories include an extra stainless steel support rod, two choices of clamps, and stainless steel stabilizer bolts that are threaded to fit the extra holes in the base of the support stand. The stainless steel support stand clamp (58950-660) is designed with a precision fit to accept a 13mm (1/2") diameter support shaft and a 16mm (5/8") diameter support rod on a support stand. Heavy-duty support clamp (14010-000) is made of a strong aluminum alloy and is fitted with oversized knobs for very secure positioning. It supports all shafts from 6 to 24mm (0.24 to 0.94") in diameter and supports rods on a 10.2 cm (4") long surface to avoid wobble and vibration.
Includes support rod.
Ordering information: Assembly of support stand (base and one support rod) required. Additional support rods, clamps, and stabilizer bolts are ordered separately.