Blowers for Laboratory Hoods, Labconco®
Supplier: Labconco
71810003726 7182200 706920010814 706840010814 71816003726 17725110010814 706940010814 71814003726 706860010814 71800003726 71808003726 17725120010814 7180400 706880010814 17725010010814 71812003726 7180600 17705110010814 706800010814 706960010814 7182000 706820010814 71802003726 706900010814
77296-336EA 13082.4 CAD
77296-336 77296-334 82006-790 82006-792 77296-332 82006-780 77296-330 82006-754 82006-786 82006-742 82006-756 82006-778 82006-744 82006-788 82006-794 82006-750 97019-806 82006-782 82006-796 82006-752 82006-784 82006-740 82006-746 82006-748
Blowers for Laboratory Hoods, Labconco®
Ventilators
Fiberglass blowers are recommended for fume hood exhaust systems in moderate to highly corrosive conditions
Coated steel blowers are recommended for low to moderately corrosive applications and have a phenolic coating on impeller and housing.
Both blowers have a dry powder epoxy-coated base and weather cover for protection in extreme weather. Sheaves are adjustable to allow for air volume adjustment. Belt driven models use a self-adjusting gravity belt tightener that ensures consistent performance because belt is held taut by weight of motor.
The housing is made from molded fiberglass-reinforced polyester for durability and long life. Impellers are made of injection-molded polypropylene to resist corrosive atmospheres.
Spectrum Blowers do not use belts, therefore use less energy and are suitable for all applications.
Ordering information: Items indicated below as REDISHIP units can be shipped within 48 hours of receiving your order.