Synthware Vacuum Adapters, Kemtech America
Supplier: Kemtech America
A741420 A741922 A742440 A741420G A741922G A742440G
60000-342EA 145.06 CAD
60000-342 60000-344 60000-346 60000-350 60000-352 60000-354
Synthware Vacuum Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Vacuum adapter is designed for evacuation or purging.
  • [ST] joint
  • PTFE or glass stopcock
Features a 2mm straight-bore PTFE or glass stopcock.