Manual Duct Dampers, Labconco
Supplier: Labconco
4741300 5983400 5981200 4726400 4724200
82006-948EA 1216.6 CAD
82006-948 82006-950 82006-952 82006-954 82010-726
Manual Duct Dampers, Labconco
Ducting Supplies
Damper fitting allows airflow balance
The duct dampers can be used with exhaust and auxiliary air ducts, and are usually placed directly above the fume hood.