VWR® Magnetic Rare Earth Stir Bars
Supplier: VWR International
001.2610.RE 001.2615.RE 001.2625.RE 001.2650.RE 001.2670.RE
89030-550PK 167.9 CAD
89030-550 89030-552 89030-554 89030-556 89030-558
VWR® Magnetic Rare Earth Stir Bars
Stirrers Magnetic Stirring Bars
Molded in pure, FDA-approved PTFE, these samarium-cobalt elliptical magnets feature increased magnetic strength and are almost totally resistant to demagnetization.
  • Autoclavable
  • PTFE construction
  • Highly resistant to demagnetization
Rare Earth magnets are identified by a chemically inert carbon black spot.