VWR® Spinbar® Magnetic Stir Bars, Saturn
Supplier: VWR International
89030-912 89030-914
89030-912CS 1441.8 CAD
89030-912 89030-914
VWR® Spinbar® Magnetic Stir Bars, Saturn
Stirrers Magnetic Stirring Bars
Many magnetic stirring bars can be impeded by powders in a liquid, but the Saturn Spinbar® Magnetic Stirring Bar rises to this stirring challenge.
  • Large sphere at the bar’s center lifts the stirring bar ends while spinning
  • Reduced surface contact area allows the bar to spin with decreased drag
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable at 121°C (250°F)
Magnetic stir bars are available with a length of 40 or 50mm (1.5 or 2") to match with a wide variety of vessels with both round and flat bottoms.