VWR® Magnetic Stir Bar Retrievers, PTFE
Supplier: VWR International
004.450 004.250.RE 004.350.RE 004.450.RE 004.250 004.350 004.150
89026-264EA 157.56 CAD
89026-264 89026-254 89026-256 89026-258 89026-260 89026-262 89026-266
VWR® Magnetic Stir Bar Retrievers, PTFE
Stirrers Magnetic Stirring Bars
Molded in pure, FDA-approved PTFE, these retrievers are inert and non-contaminating.
  • Inert
  • PTFE fabrication
  • Autoclavable
  • Magnetic cores available in different variants
Available with standard Alnico V or samarium cobalt rare earth magnetic core. Autoclavable.