Akro-Carts and Tilt Trucks, Akro-Mils™
Supplier: Akro-Mils
76590GREY 76491GREY 76490GREY 76460GREY 76461GREY 77405GREY 77410GREY 77510GREY 77610GREY 77505GREY
62661-554EA 5680.84 CAD
62661-554 62661-558 62661-560 62661-564 62661-566 62661-568 62661-570 62661-574 62661-576 62661-572
Akro-Carts and Tilt Trucks, Akro-Mils™
Trolleys
Nontipping Akro-Carts are ideal for large-sized, mediumweight jobs up to 204.1kg (450lbs.)
Akro-Tilt trucks, with their lightweight, maneuverable, and ergonomic design, make it easy for one person to handle heavyweight jobs up to 907kg (2000lbs.). Front caster wheel has roller bearings for an extra measure of stability when pushing, pulling, or turning. Large, comfortable grab handle is arc welded and reinforced for optimal durability. Full, wraparound tubular frame is constructed of 2.5cm (1") powder-coated steel for enhanced appearance and rust protection.
They feature a polyethylene container, 16-gauge zinc-plated steel tubing frame (heavy-duty model 62661-554 has powder-coated steel tubing frame), and latch bars that extend around the frame for added strength. Medium-duty models have riveted joints and the heavy-duty model has welded joints. Available with a piano-hinged lid that stays propped open during use.
Ordering information: Akro-Carts are available in gray (black lid), blue (blue lid), and green (green lid); specify color when ordering. Akro-Tilt trucks are available in gray, red, yellow, or blue; specify color when ordering. For custom colors of Akro-Lift, contact your Avantor sales representative. Specify electrical requirements of Akro-Lift when ordering.