VWR® Polygon Spinbar Magnetic Stirring Bars
Supplier: VWR International
74950-286 74950-290 74950-292 74950-294 74950-296 74950-298 74950-284 74950-288 74950-300
74950-286EA 15.07 CAD
74950-286 74950-290 74950-292 74950-294 74950-296 74950-298 74950-284 74950-288 74950-300
VWR® Polygon Spinbar Magnetic Stirring Bars
Stirrers Magnetic Stirring Bars
These VWR® stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminishe
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave-safe