VWR® Spinbar® Disposable Stir Bars, Octagon
Supplier: VWR International
58947-144 58947-146 58947-140 58947-142
58947-144PK 513.42 CAD
58947-144 58947-146 58947-140 58947-142
VWR® Spinbar® Disposable Stir Bars, Octagon
Stirrers Magnetic Stirring Bars
The multifaceted stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Ideal for single use applications such as mixing radioactive reagents
  • Disposable; eliminates potential for cross-contamination
  • Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert
  • Autoclavable
For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.