VWR® Spinplus® Magnetic Stir Bars
Supplier: VWR International
58947-820 58947-822 58947-824 58947-826 58947-828 58947-830
58947-820CS 1430.73 CAD
58947-820 58947-822 58947-824 58947-826 58947-828 58947-830
VWR® Spinplus® Magnetic Stir Bars
Stirrers Magnetic Stirring Bars
Spinplus® magnetic stirring bars add speed and efficiency to mixing operations.
  • The “+” shape creates a deep vortex and provides stable, quiet operation
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating is durable and inert.
Ordering information: Order each size separately or choose a five-piece assortment (one of each size).