VWR® Spinvane® Stir Bars for Test Tubes, Micro Vials, and Conical Bottom Centrifuge Tubes
Supplier: VWR International
55999-000 58949-270 58949-272 58949-274 58949-276
55999-000PK 1445.51 CAD
55999-000 58949-270 58949-272 58949-274 58949-276
VWR® Spinvane® Stir Bars for Test Tubes, Micro Vials, and Conical Bottom Centrifuge Tubes
Stirrers Magnetic Stirring Bars
Solid Teflon® vane has a cross-mounted, molded-in magnet that can be trimmed to fit if needed without affecting the magnet.
  • Half-round shape is designed for 16 mm O.D. test tubes
  • Half-round tapered shape is designed for conical-bottom centrifuge tubes
  • Triangular shape is designed for small conical-shaped tubes and micro vials
  • Autoclavable
Each size is manufactured for a specific size tube; can be trimmed to fit if needed without affecting the magnet.