VWR® Spinfin® Double Stir Bars
Supplier: VWR International
58948-998 58949-000 58949-004
58948-998CS 1732.92 CAD
58948-998 58949-000 58949-004
VWR® Spinfin® Double Stir Bars
Stirrers Magnetic Stirring Bars
Double Spinfin® Magnetic Stirring Bars agitate liquids in round bottom vessels such as test tubes, cylinders, and flasks, where other stirring bars cannot be used.
  • Fins on top and bottom ensure complete agitation
  • Maintains its position in a vessel and will not wobble
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.