VWR® Spinfin® Magnetic Stir Bars
Supplier: VWR International
58948-990 58948-992 58948-994 58948-996 76001-868 76001-870
58948-990CS 1275.85 CAD
58948-990 58948-992 58948-994 58948-996 76001-868 76001-870
VWR® Spinfin® Magnetic Stir Bars
Stirrers Magnetic Stirring Bars
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used
  • Maintains its position in a vessel and will not wobble
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.