SP Bel-Art Giant Polygon Spinbar Magnetic Stirring Bars without Pivot Ring, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37118-0004 F37118-0002 F37118-0006
58948-972CS 2900.21 CAD
58948-972 58948-974 58948-978
SP Bel-Art Giant Polygon Spinbar Magnetic Stirring Bars without Pivot Ring, Bel-Art Products, a part of SP
Heavy duty bars for use in stirring large volumes of fluid in vessels on large magnetic stirring machines such as the Cool Stir Large Volume Stirrer, 89009-206 or other large machines
  • Will fit into vessels with a minimum neck diameter of 32 mm
  • Multifaceted surface adds turbulence relative to similar size smooth cylindrical stirring bars
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert.