Rotors for Allegra® X-22 and Allegra X-22R Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
392271 392274 361171
CABK392243EA 3164.33 CAD
CABK392243 CABK392271 CABK392274 CABK361171
Rotors for Allegra® X-22 and Allegra X-22R Centrifuges, Beckman Coulter®
Centrifuges Rotors
These rugged rotors are constructed from heavy-duty aluminum and are engineered to provide quiet, dependable service.
  • Application: Pelleting Subcellular Organelles, Nucleic acids, Viruses, Bacteria, and More
  • Material: Heavy-Duty Aluminum
  • Quiet Process
  • Bio-Certified
  • Fixed, 45⁰ Angle
Fixed-angle rotors run at higher speeds for small volumes; swinging bucket rotors process large volumes at slower speeds.
Rotors accommodate tubes and bottles from 1.5 to 250 mL, up to six standard microplates, or two deep-well plates with a 96-well format. Rotors can be autoclaved or chemically sterilized.
Ordering information: Rotors are supplied complete and ready to use. Bottles and tubes are available separately. For bottle and tube adapters, see BK392261, BK361247, and BK392270 series.