Microfuge® 16 Microcentrifuge, Beckman Coulter®
Supplier: Beckman Coulter
Microfuge®
A46472 A46472 A46474
CABKA46472EA 6342.04 CAD
BKA46474 BKA46472 CABKA46472
Microfuge® 16 Microcentrifuge, Beckman Coulter®
Centrifuges Microcentrifuges
This non-refrigerated, cold-room compatible, high-performance microcentrifuge is ideal for fast pelleting or isolation of DNA, RNA, proteins, and viruses.
  • Brushless-Induction Drive
  • Automatic, Motorized Closure System
  • Air-Cooled Rotor Bowl
  • Polypropylene Rotors
  • Polypropylene, Snap-On Lid for Rotors
The compact Microfuge® microcentrifuge features a brushless-induction drive, 13-second acceleration and deceleration to/from maximum speed, an automatic, motorized closure system with a safety auto-lock, an air-cooled rotor bowl, a large display with push-button entry, and an automatic lid release upon completion of sample runs.
Runs may be set to pulse, timed (one-second increments), or continuous, while speed may be set to standard or soft start/stop in 100 rpm increments. Polypropylene rotors are autoclavable and supplied with a polypropylene, snap-on lid.
Certifications: UL listed. C-UL Listed. CE marked.