VWR® Incubating Microplate Shaker
Supplier: VWR
VWR® symphony™
980140PL-C 980140-C 980140PL 980140
10027-132EA 9785.45 CAD
10027-132 97043-606 10027-128 12620-930
VWR® Incubating Microplate Shaker
Shakers Shaking Incubators
Incubating microplate shakers are ideal for cell cultures, hybridizations, and suspensions.
  • Optimized for shaking microplates or microtubes
  • Digital display for speed, temperature, and time
  • Microprocessor control
  • Timer with audible alarm
  • Available with opaque lid for light-sensitive samples
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Microprocessor PID temperature controller maintains precise temperature control from 5° above ambient to 65 °C (149 °F)
The triple eccentric drive features permanently lubricated ball bearings. The maintenance-free, brushless DC motor provides a long service life. A Lexan® polycarbonate lid permits viewing of samples from all sides without disturbing internal temperature. Platform dimensions: 19.7Wx27.9Dcm (73/4x11").
Optional microtube rack attaches to platform to hold 1.5 ml or 2 ml microtubes and has a capacity of 70 tubes. Units with opaque lid are designed for light sensitive samples.
Easy-to-use controls allow user to adjust temperatures in 1 °C increments. Built-in timer with LED display will show elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero. Holds four microplates or two microtube racks.
Certifications: RoHS certified. TUV Listed, CE Marked. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a 2.3 m (7.5') detachable, three-wire cord and plug. Supplied without accessories which must be ordered separately. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.