VWR® Microplate Shaker
Supplier: VWR International
12620-926 10027-120
12620-926EA 1595.84 CAD
12620-926 10027-120
VWR® Microplate Shaker
Shakers Microplate Shakers
Ideal for immunoassays and general microplate shaking, microplate shakers feature a four-place platform that holds either two or four microplates or deep well blocks.
  • Optimized for shaking microplates
  • Digital display for speed and time
  • Microprocessor control
  • Timer with audible alarm
  • Holds 2 or 4 microplates
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Plates are firmly locked into place with retaining springs. Microprocessor variable speed control provides consistent, uniform mixing action. Ramping feature slowly increases speed to set point for added safety. Independent LED displays feature speed and time. Timer will display elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero. The triple eccentric drive features permanently lubricated ball bearings. The maintenance-free, brushless DC motor provides a long service life. Ideal for use in incubators, CO2 incubators, or cold rooms; shaker accommodates a -10 to 60 °C (-14 to 140°F), 80% relative humidity, non-condensing environment. Durable base is constructed of cast aluminum for extra stability.
Certifications: RoHS certified. CSA listed and CE Marked. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Units include a 2.3m (7.5') detachable, three-wire cord and plug. Optional microtube rack attaches to platform to hold 1.5 mL or 2 mL microtubes and has a capacity of 70 tubes. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.